Order Entry
Netherlands
ContactUsLinkComponent
 
 
4'-(Methoxycarbonyl)benzo-15-crown-5 (4'-(Methoxycarbonyl)benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥96.0% (by GC)
  TCIAM1489-1G
 :  TCI
 :  
undefined
 
4'-(Methoxycarbonyl)benzo-15-crown-5 (4'-(Methoxycarbonyl)benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥96.0% (by GC)
  TCIAM1489-1G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Solubility in Methanol:
    almost transparency

 

 

 :
 :82°C
 :GHS07
 :56683-56-8
 :C₁₆H₂₂O₇
 :MFCD06797129