
Ethyl-N-hydroxyacetimidate ≥95.0% (by GC)
: TCI
:10576-12-2
:
:C₄H₉NO₂
:MFCD00002114
:103.12 g/mol
:60°C
:234-165-7
:GHS07
:Refrigerator
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together