(4-Nitrophenyl)-α-D-galactopyranoside ≥98.0% (by HPLC) substrate for alpha-D-Galactosidase
Supplier: TCI
Certificates
About this item
CAS Number:7493-95-0
:
Formula:C₁₂H₁₅NO₈
MDL Number:MFCD00065050
:301.26 g/mol
Storage Temperature:Refrigerator
EINECS:231-343-6
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:230 to 246 deg (C=1, H2O)
- alpha-Galactosidase detection test:to pass test
Specification Test Results
Frequently Bought Together