Order Entry
Netherlands
ContactUsLinkComponent
6-Amino-1,4-benzodioxane ≥98.0% (by GC, titration analysis)
Irritant
6-Amino-1,4-benzodioxane ≥98.0% (by GC, titration analysis)
Catalog # TCIAE0385-25ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
6-Amino-1,4-benzodioxane ≥98.0% (by GC, titration analysis)
Catalog # TCIAE0385-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :151.17 g/mol
Boiling Pt:162°C
EINECS:244-718-4
ADR:6.1,III
 :GHS07
CAS Number:22013-33-8
Formula:C₈H₉NO₂
MDL Number:MFCD00006824