
3-(Trimethoxysilyl)propyl methacrylate ≥98.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:2530-85-0
:
Formula:C₁₀H₂₀O₅Si
MDL Number:MFCD00008593
:248.35 g/mol
Boiling Pt:110°C
Flash Pt:92°C
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:219-785-8
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together