


3,6-Dioxaoctamethylenediamine ≥98.0% (by GC, titration analysis)
: TCI
:929-59-9
:
:C₆H₁₆N₂O₂
:MFCD00040474
:148.20 g/mol
:123°C
:121°C
:213-203-6
:2735
:8,II
:GHS05
:GHS07
:GHS08
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Frequently Bought Together