Order Entry
Netherlands
ContactUsLinkComponent
4-(4-Bromophenyl)piperidin-4-ol ≥98.0% (by GC, titration analysis)
Irritant
4-(4-Bromophenyl)piperidin-4-ol ≥98.0% (by GC, titration analysis)
Catalog # TCIAB1719-10G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-(4-Bromophenyl)piperidin-4-ol ≥98.0% (by GC, titration analysis)
Catalog # TCIAB1719-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    165.0 to 169.0 deg-C

Specification Test Results

About this item

 :
 :256.14 g/mol
EINECS:261-065-0
 :GHS07
CAS Number:57988-58-6
Formula:C₁₁H₁₄BrNO
MDL Number:MFCD00067034