
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) ≥98.0% (by GC, titration analysis) catalyst for oxidation free radical
: TCI
:95407-69-5
:
:C₁₀H₂₀NO₂
:MFCD00270334
:186.27 g/mol
:GHS07
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Iodometric Titration):min. 98.0 %
- Melting point:40.0 to 45.0 deg-C
Frequently Bought Together