
1,1-Dimethylguanidinium sulphate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:598-65-2
:
Formula:C₆H₂₀N₆O₄S
MDL Number:MFCD00013131
:272.33 g/mol
EINECS:209-946-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together