

1-Methyl-3-nitroguanidine ≥98.0% (by titrimetric analysis) wetted with ca. 25% water (unit weight on dry weight basis)
Catalog # TCIAM0749-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 % (after drying)
- Melting point:158.0 to 162.0 deg-C
- Solubility in hot Water:very faint turbidity
- Water:25.0 and le 35.0 %
Specification Test Results
About this item
:
:147.09 g/mol
ADR:4.1,III
:Moistened with water
UN:1325
:GHS02
:GHS07
CAS Number:4245-76-5
Formula:C₂H₅N₅O₃
MDL Number:MFCD00024167