Order Entry
Netherlands
ContactUsLinkComponent
3-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
Irritant
3-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAM1793-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAM1793-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal

Specification Test Results

About this item

 :
 :168.02 g/mol
 :GHS07
CAS Number:128312-11-8
Formula:C₇H₉BO₂S
MDL Number:MFCD01319105