
4-Iodophenol ≥98.0% (by GC) for biochemical research
: TCI
:540-38-5
:
:C₆H₅IO
:MFCD00002327
:220.01 g/mol
:14,05036
:287 °C (1013 hPa)
:92…93 °C
:2,13 g/cm³ (25 °C)
:Ambient
:208-745-5
:8,III
:GHS07
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:90.0 to 94.0 deg-C
- Chemiluminescence test (Luminol/POD/H2O2):to pass test
Frequently Bought Together