
2-Hydroxy-4-(trifluoromethyl)pyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:50650-59-4
:
Formula:C₆H₄F₃NO
MDL Number:MFCD00153195
:163.1 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:156.0 to 162.0 deg-C
Specification Test Results
Frequently Bought Together