Order Entry
Netherlands
ContactUsLinkComponent
4-Chloro-1-butanol ≥85.0% (by GC) (contains varying amounts of Tetrahydrofuran)
Flammable
Irritant
Health
4-Chloro-1-butanol ≥85.0% (by GC) (contains varying amounts of Tetrahydrofuran)
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
Health
4-Chloro-1-butanol ≥85.0% (by GC) (contains varying amounts of Tetrahydrofuran)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 85.0 %

Specification Test Results

Frequently Bought Together