
(1S)-(+)-Camphor-10-sulphonyl chloride ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:21286-54-4
:
Formula:C₁₀H₁₅ClO₃S
MDL Number:MFCD00064156
:250.75
Melting Pt:70 °C
EINECS:244-314-8
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
- Melting point:65.0 to 69.0 deg-C
- Specific rotation [a]20/D:+31.0 to +34.0 deg (C=1, CHCl3)
Specification Test Results
Related Information
Frequently Bought Together