
(1R)-(-)-Camphor-10-sulphonyl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:39262-22-1
:
Formula:C₁₀H₁₅ClO₃S
MDL Number:MFCD00064155
Melting Pt:64…67 °C
UN:3261
ADR:8,III
:250.75
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:66.0 to 68.0 deg-C
- Specific rotation [a]20/D:-26.0 to -33.0 deg (C=1, CHCl3)
- Solubility in Toluene:almost transparency
Specification Test Results
Related Information
Frequently Bought Together