

2-Hexene cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAH1009-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
Specification Test Results
About this item
:
:84.16 g/mol
Boiling Pt:67 °C (1013 hPa)
Melting Pt:–98.5 °C
Density:0,678 g/cm³ (20 °C)
Flash Pt:-20°C
Flash Pt:–20°C
Storage Temperature:Ambient
EINECS:209-755-2
UN:3295
ADR:3,II
:GHS02
:GHS08
CAS Number:592-43-8
Formula:C₆H₁₂
MDL Number:MFCD00009473