
2,5-Diphenyloxazole ≥99.0% (by HPLC, total nitrogen) for scintillation spectrometry
: TCI
:92-71-7
:
:C₁₅H₁₁NO
:MFCD00005306
:221.26 g/mol
:360 °C (1013 hPa)
:70…72 °C
:1,094 g/cm³ (99 °C)
:Ambient
:202-181-3
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (with Total Nitrogen):min. 99.0 %
- Melting point:71.0 to 74.0 deg-C
- Solubility in Methanol:very faint turbidity
Frequently Bought Together