

1,4-Dihydroxy-9,10-anthraquinone ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:81-64-1
:
Formula:C₁₄H₈O₄
MDL Number:MFCD00001209
:240.22 g/mol
Merck Index:14,08064
Boiling Pt:465 °C (1013 hPa)
Melting Pt:202 °C
Density:1,278 g/cm³ (20 °C)
EINECS:201-368-7
:GHS07
:GHS08
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:196.0 to 202.0 deg-C
Specification Test Results
Frequently Bought Together