
2,6-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:73852-17-2
:
Formula:C₆H₅BCl₂O₂
MDL Number:MFCD00064869
:190.82 g/mol
Melting Pt:151°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):96.0 to 111.0 %
Specification Test Results
Related Information
Frequently Bought Together