

Dipropyl ether ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:111-43-3
:
Formula:C₆H₁₄O
MDL Number:MFCD00009376
:102.18 g/mol
Merck Index:14,07857
Boiling Pt:90°C
Melting Pt:–122 °C
Density:0,736 g/cm³ (20 °C)
Flash Pt:21°C
Storage Temperature:Ambient
EINECS:203-869-6
UN:2384
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together