Order Entry
Netherlands
ContactUsLinkComponent
 
 
(1-Pentyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
  TCIAA0862-25G
 :  TCI
 :  
undefined
 
(1-Pentyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
  TCIAA0862-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    164.0 to 168.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

 

 :
 :413.34 g/mol
 :168 °C
 :244-374-5
 :GHS07
 :21406-61-1
 :H₃C(CH₂)₄P(C₆H₅)₃Br
 :MFCD00031630