Order Entry
Netherlands
ContactUsLinkComponent
3-(Trifluoromethoxy)phenylboronic acid (contains varying amounts of Anhydride)
Irritant
3-(Trifluoromethoxy)phenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAT2362-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3-(Trifluoromethoxy)phenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAT2362-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Neutralization titration):
    97.0 to 110.0 %

Specification Test Results

About this item

 :
 :205.93 g/mol
 :GHS07
CAS Number:179113-90-7
Formula:C₇H₆BF₃O₃
MDL Number:MFCD01320697