
2-Cyanophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:138642-62-3
:
Formula:C₇H₆BNO₂
MDL Number:MFCD01632208
:146.94 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 114.0 %
Specification Test Results
Related Information
Frequently Bought Together