

2,6-Dinitro-4-methylanisole ≥98.0% (by GC)
Catalog # TCIAM1842-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:122.0 to 126.0 deg-C
Specification Test Results
About this item
:
UN:2930
ADR:6.1,II
:GHS02
:GHS06
CAS Number:29455-11-6
Formula:C₈H₈N₂O₅
MDL Number:MFCD00192097