Order Entry
Netherlands
ContactUsLinkComponent
2,6-Diamino-9,10-anthraquinone ≥97.0% (by total nitrogen basis)
Irritant
2,6-Diamino-9,10-anthraquinone ≥97.0% (by total nitrogen basis)
Catalog # TCIAD3180-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,6-Diamino-9,10-anthraquinone ≥97.0% (by total nitrogen basis)
Catalog # TCIAD3180-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Yellow to Amber to Dark red powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %

Specification Test Results

About this item

 :
 :238.2414 g/mol
Melting Pt:320°C
EINECS:205-013-7
 :GHS07
CAS Number:131-14-6
Formula:C₁₄H₁₀N₂O₂
MDL Number:MFCD00001234