Order Entry
Netherlands
ContactUsLinkComponent
3-Bromo-2-nitroanisole ≥98.0% (by GC)
3-Bromo-2-nitroanisole ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
3-Bromo-2-nitroanisole ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    72.0 to 76.0 deg-C

Specification Test Results

Frequently Bought Together