4-Methoxy-N-methylaniline ≥96.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5961-59-1
:
Formula:C₈H₁₁NO
MDL Number:MFCD00008399
:137.18 g/mol
Boiling Pt:136°C
EINECS:227-735-1
Specification Test Results
- Appearance:White or Colorless to Brown powder to lump to clear liquid
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
- Melting point:33.0 to 37.0 deg-C
Specification Test Results
Frequently Bought Together