Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
4-Methyl-1H-pyrazole 97%
 
undefined
 
 
4-Methyl-1H-pyrazole 97%

 

  • Appearance:
    Clear colorless to yellow Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4920 to 1.4980 (20°C, 589 nm)
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together