Order Entry
Netherlands
ContactUsLinkComponent
Tetrabutylammonium hydroxide 10% in methanol
Flammable
Corrosive
Irritant
Health
Tetrabutylammonium hydroxide 10% in methanol
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Tetrabutylammonium hydroxide 10% in methanol
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Concentration (Neutralization titration):
    10.0 to 11.0 w/w%
  • Haloid:
    max. 0.5 %

Specification Test Results

Frequently Bought Together