
Tetrabutylammonium hydroxide 10% in water for ion pair chromatography
: TCI
:2052-49-5
:
:C₁₆H₃₇NO
:MFCD00009425
:259.48 g/mol
:Refrigerator
:218-147-6
:3267
:8,II
:GHS05
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
- Absorbance (230nm):max. 0.100
- Absorbance (254nm):max. 0.050
- Absorbance (400nm):max. 0.010
Frequently Bought Together