

2-Isopropoxy-4,4,6-trimethyl-1,3,2-dioxaborinane ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:61676-61-7
:
Formula:C₉H₁₉BO₃
MDL Number:MFCD15143641
:186.06 g/mol
Boiling Pt:59°C
Flash Pt:49°C
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together