


Bromomethyl methyl ether ≥95.0% (by GC)
: TCI
:13057-17-5
:
:C₂H₅BrO
:MFCD00000171
:124.97 g/mol
:88°C
:26°C
:1992
:3,III
:GHS02
:GHS06
:GHS08
:Refrigerator
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together