
2-Mercaptopyridine-N-oxide ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1121-31-9
:
Formula:C₅H₅NOS
MDL Number:MFCD00006196
:127.17 g/mol
EINECS:214-329-4
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Iodometric Titration):min. 95.0 %
- Melting point:70.0 to 73.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together