4,4'-Anisoin ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:119-52-8
:
Formula:C₁₆H₁₆O₄
MDL Number:MFCD00008411
:272.30 g/mol
EINECS:204-330-8
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:110.0 to 113.0 deg-C
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together