
4-Methyl-3-nitrophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:80500-27-2
:
Formula:C₇H₈BNO₄
MDL Number:MFCD00191550
:180.96 g/mol
:GHS07
Specification Test Results
- Purity (Neutralization titration):97.0 to 111.0 %
Specification Test Results
Related Information
Frequently Bought Together