Pterostilbene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:537-42-8
:
Formula:C₁₆H₁₆O₃
MDL Number:MFCD00238710
:256.2964 g/mol
Storage Temperature:Koelkast
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together