3-Methoxy-N,N-dimethylaniline ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15799-79-8
:
Formula:C₉H₁₃NO
MDL Number:MFCD00051779
:151.21 g/mol
Boiling Pt:123°C
ADR:6.1,III
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together