
1H-Pyrazole-4-carboxylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:37718-11-9
:
Formula:C₄H₄N₂O₂
MDL Number:MFCD00077436
:112.09 g/mol
Melting Pt:275°C
EINECS:253-641-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together