
1H-Pyrazole-3-carboxylic acid ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1621-91-6
:
Formula:C₄H₄N₂O₂
MDL Number:MFCD00077436
:112.09 g/mol
Melting Pt:213°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Frequently Bought Together