

TATU (O-(7-aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:873798-09-5
:
Formula:C₁₀H₁₅BF₄N₆O
MDL Number:MFCD08064301
ADR:8,III
:322.1 g/mol
UN:1759
:GHS05
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together