Order Entry
Netherlands
ContactUsLinkComponent
Pesticide reference standard, Chlordane 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, Chlordane 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL11200000CY
 :  
Pesticide reference standard, Chlordane 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL11200000CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    57-74-9
  • Description:
    Chlordane 10 µg/ml in cyclohexane
  • Formula:
    C₁₀H₆Cl₈
  • Description:
    Chlordane