Order Entry
Netherlands
ContactUsLinkComponent
Disinfectant surface wipes, Distel
Disinfectant surface wipes, Distel
Catalog # 115-1770
Supplier:  TRISTEL SOLUTIONS
undefined
Disinfectant surface wipes, Distel
Catalog # 115-1770
Supplier:  TRISTEL SOLUTIONS

Specifications

  • Packed:
    Sachet of 250 wipes
  • Description:
    Surface disinfectant wipe, 1:100, non-fragranced sachet
  • L×W:
    155×110 mm
  • Seq:
    01
  • Pk:
    250

Specifications

About this item

The formulation of Distel wipes is based on a unique carrier technology enabling the active ingredients to be carried rapidly through cell walls of microorganisms. The synergistic blend of active ingredients produces the fastest acting and most efficient virucidal and sporicidal disinfectant available in non-oxidising technology. This allows both rapid and effective results, and makes it safer to use than conventional products.

  • Fast acting, simple and effective disinfectant
  • Colourless and odourless
  • Shelf life for tubs: 2 years; for individual sachets: 3 years
  • Active constituents are approved under European Biocidal Products Directive (ref 98/8/EC) and REACH compliance
  • Conforms to the medical device directive 93/42/EEC and 2007/47/EEC amendments

Distel wipes are suitable for use on most hard surfaces in high risk areas to destroy bacteria, viruses, fungi and mycobacteria and to denature DNA/RNA. Intended for use on hard surfaces of non-invasive, non-lumened medical devices such as monitors, IV poles, beds, patient trolleys, ultrasound and X-ray equipment.

Concentrate dilution 1:100: Wipes are suitable for heavily soiled and/or high risk areas.
Concentrate dilution 1:10: Wipes can be used in the presence of blood, body spills and bio-hazards.

Bactericidal activity: Pseudomonas aeruginosa, Campylobacter spp, Staphylococcus spp, Streptococcus spp, Leptospira spp, Escherichia coli, Enterococcus spp , Klebsiella spp, Salmonella spp, Listeria monocytogenes, Legionella spp, Proteus vulgaris, Rhodococcus spp, Serratia spp, Shigella spp, Helicobacter pylori, MRSA, Mycobacterium tuberculosis, Mycobacterium terrae, Mycobacterium avium.

Virucidal activity: Coronavirus (SARS), Adenovirus type 5, Influenza, H5NI

Ordering information: Available in 100 or 200 wipe tubs, or in individual sachet format.
Caution: Use biocides safely. Always read the label and product information before use.

*The product(s) on this page are outside the scope of the EU Directive 2019/904