Temperature probe: Pt1000 Body material: glass Body length: 170 mm Body diameter: 12 mm Electrolyte: 3 mol/l KCl pH measurement range: 0 to 14 Temperature range: -5 ... +100 °C Weight 1,04 KG 1 * 1 items
Catalog # KNIC78870
Supplier: KNICK