ContactUsLinkComponent
Staphylococcus aureus subsp. aureus WDCM 00032 Vitroids 50,000-150,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000327-10EA
Supplier:  Merck
New Product
Staphylococcus aureus subsp. aureus WDCM 00032 Vitroids 50,000-150,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000327-10EA
Supplier:  Merck