Salmonella enterica subsp. enterica serovar Enteritidis WDCM 00030 Vitroids 50,000-150,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000307-10EA
Supplier: Merck