ContactUsLinkComponent
Escherichia coli WDCM 00013 Vitroids 15-80 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000132-10EA
Supplier:  Merck
New Product
Escherichia coli WDCM 00013 Vitroids 15-80 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000132-10EA
Supplier:  Merck