ContactUsLinkComponent
Burkholderia cepacia CECT 4137 Vitroids 15-80 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT041372-10EA
Supplier:  Merck
New Product
Burkholderia cepacia CECT 4137 Vitroids 15-80 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT041372-10EA
Supplier:  Merck