ContactUsLinkComponent
Bacillus cereus WDCM 00001 Vitroids 1,000-10,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000016-10EA
Supplier:  Merck
New Product
Bacillus cereus WDCM 00001 Vitroids 1,000-10,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000016-10EA
Supplier:  Merck