ContactUsLinkComponent
Alicyclobacillus acidoterrestris CECT 7094 Vitroids 80-130 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT070943-10EA
Supplier:  Merck
New Product
Alicyclobacillus acidoterrestris CECT 7094 Vitroids 80-130 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT070943-10EA
Supplier:  Merck